ContactUsLinkComponent
L-Cysteine hydrochloride salt is routinely used with Ellman's Reagent assays as a sulfhydryl standard. In addition, it is also used as a supplement for protein refolding experiments. 1 * 5 g
Catalog # GENO786-713
Supplier:  G-Biosciences
L-Cysteine hydrochloride salt is routinely used with Ellman's Reagent assays as a sulfhydryl standard. In addition, it is also used as a supplement for protein refolding experiments. 1 * 5 g
Catalog # GENO786-713
Supplier:  G-Biosciences
Frequently Bought Together