ContactUsLinkComponent
Irreversible inhibitor of serine proteases, including trypsin & chymotrypsin null
Catalog # GENO786-787
Supplier:  G-Biosciences
Irreversible inhibitor of serine proteases, including trypsin & chymotrypsin null
Catalog # GENO786-787
Supplier:  G-Biosciences