ContactUsLinkComponent
-Aminobenzamidine Agarose primary application is for the removal and/or purification of trypsin-like proteases. -aminobenzamidin e (PAB) is a synthetic inhibitor of trypsin-like proteases and has been covalently coupled to 6% crosslinked agarose. 1 * 25 mL
  GENO786-692
-Aminobenzamidine Agarose primary application is for the removal and/or purification of trypsin-like proteases. -aminobenzamidin e (PAB) is a synthetic inhibitor of trypsin-like proteases and has been covalently coupled to 6% crosslinked agarose. 1 * 25 mL
  GENO786-692