ContactUsLinkComponent
Spectinomycin is an aminocyclitol antibiotic which acts by binding to the 30S subunit of the bacterial ribosome and inhibiting prote in synthesis. Its activity is generally modest, particularly against gram-positive bacteria. Some gram-negative bacte 1 * 25 g
Catalog # GENORC-194
Supplier:  G-Biosciences
Spectinomycin is an aminocyclitol antibiotic which acts by binding to the 30S subunit of the bacterial ribosome and inhibiting prote in synthesis. Its activity is generally modest, particularly against gram-positive bacteria. Some gram-negative bacte 1 * 25 g
Catalog # GENORC-194
Supplier:  G-Biosciences
Frequently Bought Together