ContactUsLinkComponent
Tetracycline is a bacteriostatic antibiotic with activity against both gram-positive and gram-negative bacteria. Within the cell tet racycline binds reversibly to the 30S subunit of the ribosome, preventing the binding of aminoacyl tRNA and inhibitin 1 * 100 g
Catalog # GENORC-198
Supplier:  G-Biosciences
Tetracycline is a bacteriostatic antibiotic with activity against both gram-positive and gram-negative bacteria. Within the cell tet racycline binds reversibly to the 30S subunit of the ribosome, preventing the binding of aminoacyl tRNA and inhibitin 1 * 100 g
Catalog # GENORC-198
Supplier:  G-Biosciences
Frequently Bought Together