Ticarcillin is an inhibitor of bacterial cell wall synthesis. It inhibits the crosslinking of peptidoglycan by binding and inactivat ing of transpeptidases. High activity against gram-negative bacteria and the combination is effective against resista 1 * 10 g
GENORC-200