Silicone paste A technical grade 1 * 100 g
APPC211630.1208
Frequently Bought Together
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
Various replacement parts
: SCIENCIX
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals