Bromine (Bromate-Bromide) 0.05 mol/l (0.1N) volumetric solution 1 * 1 L
Catalog # APPC182000.1211
Supplier: PanReac AppliChem
Bromine (Bromate-Bromide) 0.05 mol/l (0.1N) volumetric solution 1 * 1 L
Catalog # APPC182000.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% unstabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), BAKER, J.T.Baker®
Supplier: Avantor
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers), technical
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals