Conductivity Standard 1413 uS/cm (25C) 1 * 250 mL
Catalog # APPC394659.1209
Supplier: PanReac AppliChem
Conductivity Standard 1413 uS/cm (25C) 1 * 250 mL
Catalog # APPC394659.1209
Supplier: PanReac AppliChem
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Quantitative Filter Papers
Supplier: VWR Collection
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Orion™ Conductivity Standards and Solutions, Thermo Scientific
Supplier: Thermo Orion