Conductivity Standard 12,88 mS/cm (25C) 1 * 250 mL
Catalog # APPC394658.1209
Supplier: PanReac AppliChem
Conductivity Standard 12,88 mS/cm (25C) 1 * 250 mL
Catalog # APPC394658.1209
Supplier: PanReac AppliChem
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW