FLUORESCENT DYE 594-I STREPTAVIDIN FLUORESCENCE DYE 594-I WITH STREPTAVIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594-I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE 1 * 200 µG
Catalog # ABNOU0291
Supplier: Abnova