FLUORESCENT DYE 570-M STREPTAVIDIN FLUORESCENCE DYE 570-M WITH STREPTAVIDIN CONJUGATE. ALL THE FLUORESCENT DYE M SERIES PRODUCTS AR E DESIGNED TO BE MAXIMALLY EXCITED BY ONE OF THE MAJOR LIGHT SOURCES EQUIPPED IN FLOW CYTOMETERS. BESIDES, IN COMPARI 1 * 100 µG
Catalog # ABNOU0290
Supplier: Abnova