FLUORESCENT DYE 510-M STREPTAVIDIN FLUORESCENCE DYE 510-M WITH STREPTAVIDIN CONJUGATE. ALL THE FLUORESCENT DYE M SERIES PRODUCTS AR E DESIGNED TO BE MAXIMALLY EXCITED BY ONE OF THE MAJOR LIGHT SOURCES EQUIPPED IN FLOW CYTOMETERS. BESIDES, IN COMPARI 1 * 100 µG
Catalog # ABNOU0282
Supplier: Abnova