FLUORESCENT DYE 405-I STREPTAVIDIN FLUORESCENCE DYE 405-I WITH STREPTAVIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENC E DYE 405-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10). FLUORESCENCE DYE 405- 1 * 200 µG
Catalog # ABNOU0277
Supplier: Abnova