ContactUsLinkComponent
FLUORESCENT DYE 350-I STREPTAVIDIN FLUORESCENCE DYE 350-I WITH STREPTAVIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENC E DYE 350-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10) 200 ug 1 * 200 µG
Catalog # ABNOU0275
Supplier:  Abnova
FLUORESCENT DYE 350-I STREPTAVIDIN FLUORESCENCE DYE 350-I WITH STREPTAVIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENC E DYE 350-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10) 200 ug 1 * 200 µG
Catalog # ABNOU0275
Supplier:  Abnova