ContactUsLinkComponent
TOTAL SUSPENDED SOLIDS 10MG/L CALIBRATION STANDARD 1 * 500 mL
Catalog # PROORTC-TSS10-500
TOTAL SUSPENDED SOLIDS 10MG/L CALIBRATION STANDARD 1 * 500 mL
Catalog # PROORTC-TSS10-500