ContactUsLinkComponent
LIVE CELL TRACKING KIT (ORANGE FLUORESCENCE) LIVE CELL TRACKING KIT (ORANGE FLUORESCENCE) IS A SET OF TOOL USED TO LABEL CELLS FOR FLUORESCENCE MICROSCOPIC AND FLOW CYTOMETRIC INVESTIGATIONS OF CELLULAR FUNCTIONS 1 KIT 1 * 1 KIT
Catalog # ABNOKA4105
Supplier:  Abnova
LIVE CELL TRACKING KIT (ORANGE FLUORESCENCE) LIVE CELL TRACKING KIT (ORANGE FLUORESCENCE) IS A SET OF TOOL USED TO LABEL CELLS FOR FLUORESCENCE MICROSCOPIC AND FLOW CYTOMETRIC INVESTIGATIONS OF CELLULAR FUNCTIONS 1 KIT 1 * 1 KIT
Catalog # ABNOKA4105
Supplier:  Abnova