ContactUsLinkComponent
Bottles glass ointment jar white flint 120ml type III soda lime glass with fitted R3/58 natural PP wadless caps max height 63.50mm max dia 62.30mm 1 * 20 items
Catalog # 215-3329
Supplier:  Azpack
undefined
Bottles glass ointment jar white flint 120ml type III soda lime glass with fitted R3/58 natural PP wadless caps max height 63.50mm max dia 62.30mm 1 * 20 items
Catalog # 215-3329
Supplier:  Azpack
Frequently Bought Together