ContactUsLinkComponent
Bottles glass ointment jar white flint 30ml type III soda lime glass with fitted R3/38 white PP wadless caps max height 46.80mm max dia 41.30mm 1 * 20 items
Catalog # 215-3319
Supplier:  Azpack
undefined
Bottles glass ointment jar white flint 30ml type III soda lime glass with fitted R3/38 white PP wadless caps max height 46.80mm max dia 41.30mm 1 * 20 items
Catalog # 215-3319
Supplier:  Azpack
Frequently Bought Together