ContactUsLinkComponent
Bottles glass ointment jar white flint 15ml type III soda lime glass with fitted R3/38 black urea pvdc woodpulp lined caps max heigh t 35.30mm max dia 41.70mm 1 * 40 items
Catalog # 215-3309
Supplier:  Azpack
undefined
Bottles glass ointment jar white flint 15ml type III soda lime glass with fitted R3/38 black urea pvdc woodpulp lined caps max heigh t 35.30mm max dia 41.70mm 1 * 40 items
Catalog # 215-3309
Supplier:  Azpack
Frequently Bought Together