ContactUsLinkComponent
Beakers 'Tri-Stir' 100ml polypropylene flat bottom 10ml graduations with V notched corners for easy pour and less spillage capability. 1 * 10 items
Catalog # 213-0403
Supplier:  Azpack
undefined
Beakers 'Tri-Stir' 100ml polypropylene flat bottom 10ml graduations with V notched corners for easy pour and less spillage capability. 1 * 10 items
Catalog # 213-0403
Supplier:  Azpack
Frequently Bought Together