ContactUsLinkComponent
Peristaltic pump tubing, PVC Solvaflex, yellow/yellow tabs, 1.42 mm id, for use with SPS 3 autosampler when pumping low volatile org anic samples, 12/pk 1 * 12 items
Catalog # HEWL3710069300
Supplier:  Agilent
Peristaltic pump tubing, PVC Solvaflex, yellow/yellow tabs, 1.42 mm id, for use with SPS 3 autosampler when pumping low volatile org anic samples, 12/pk 1 * 12 items
Catalog # HEWL3710069300
Supplier:  Agilent
Frequently Bought Together