qPCR detection of Streptococcus pneumoniae. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curv e, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-S.PNEU
Supplier: PRIMERDESIGN