ContactUsLinkComponent
qPCR detection of Hepatitis Delta Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome v iruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HDV-S
Supplier:  PRIMERDESIGN
qPCR detection of Hepatitis Delta Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome v iruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HDV-S
Supplier:  PRIMERDESIGN