ContactUsLinkComponent
qPCR detection of Human Papillomavirus 52 and 52b. Contains: Primer and probe mix, Reverse Transcription target specific primers (RN A genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HPV52
Supplier:  PRIMERDESIGN
qPCR detection of Human Papillomavirus 52 and 52b. Contains: Primer and probe mix, Reverse Transcription target specific primers (RN A genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HPV52
Supplier:  PRIMERDESIGN