ContactUsLinkComponent
qPCR detection of African Horse Sickness Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA g enome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-AHSV-
Supplier:  PRIMERDESIGN
qPCR detection of African Horse Sickness Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA g enome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-AHSV-
Supplier:  PRIMERDESIGN