qPCR detection of Aleutian Disease Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-ALDV-
Supplier: PRIMERDESIGN