ContactUsLinkComponent
qPCR detection of Budgerigar Fledgling Disease Virus (avian polyomavirus). Contains: Primer and probe mix, Reverse Transcription tar get specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-APV-S
Supplier:  PRIMERDESIGN
qPCR detection of Budgerigar Fledgling Disease Virus (avian polyomavirus). Contains: Primer and probe mix, Reverse Transcription tar get specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-APV-S
Supplier:  PRIMERDESIGN