ContactUsLinkComponent
qPCR detection of Mycobacterium Tuberculosis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cu rve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-TB
Supplier:  PRIMERDESIGN
qPCR detection of Mycobacterium Tuberculosis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cu rve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-TB
Supplier:  PRIMERDESIGN