qPCR detection of Theileria equi (formally Babesia equi). Contains: Primer and probe mix, Reverse Transcription target specific prim ers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-TH.EQ
Supplier: PRIMERDESIGN