ContactUsLinkComponent
qPCR detection of Chlamydophila pneumoniae. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.PNE
Supplier:  PRIMERDESIGN
qPCR detection of Chlamydophila pneumoniae. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.PNE
Supplier:  PRIMERDESIGN