ContactUsLinkComponent
qPCR detection of Canine parainfluenza virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA gen ome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-CPIV-
Supplier:  PRIMERDESIGN
qPCR detection of Canine parainfluenza virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA gen ome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-CPIV-
Supplier:  PRIMERDESIGN