qPCR detection of H1N1 influenza. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-H1N1-
Supplier: PRIMERDESIGN