ContactUsLinkComponent
qPCR detection of Lyme disease. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses on ly), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-LYME
Supplier:  PRIMERDESIGN
qPCR detection of Lyme disease. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses on ly), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-LYME
Supplier:  PRIMERDESIGN