qPCR detection of Mycobacterium avium subspecies paratuberculosis. Contains: Primer and probe mix, Reverse Transcription target spec ific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-MAP-S
Supplier: PRIMERDESIGN