ContactUsLinkComponent
qPCR detection of Human T-lymphotropic virus Type I. Contains: Primer and probe, RT target specific primers (RNA viruses only), Stan dard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HTLV1
Supplier:  PRIMERDESIGN
qPCR detection of Human T-lymphotropic virus Type I. Contains: Primer and probe, RT target specific primers (RNA viruses only), Stan dard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HTLV1
Supplier:  PRIMERDESIGN