ContactUsLinkComponent
qPCR detection of Avian Infectious Bronchitis Virus (IBV). Contains: Primer and probe, RT target specific primers (RNA viruses only) , Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-IBV
Supplier:  PRIMERDESIGN
qPCR detection of Avian Infectious Bronchitis Virus (IBV). Contains: Primer and probe, RT target specific primers (RNA viruses only) , Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-IBV
Supplier:  PRIMERDESIGN