qPCR detection of Human Rhinovirus Subtype 1B. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard c urve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HRV1B
Supplier: PRIMERDESIGN