ContactUsLinkComponent
qPCR detection of Human Human Rhinovirus all subtypes (generic). Contains: Primer and probe mix, Reverse Transcription target specif ic primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HRVSP
Supplier:  PRIMERDESIGN
qPCR detection of Human Human Rhinovirus all subtypes (generic). Contains: Primer and probe mix, Reverse Transcription target specif ic primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HRVSP
Supplier:  PRIMERDESIGN
Frequently Bought Together