qPCR detection of Infectious Pancreatic Necrosis Virus. Contains: Primer and probe mix, Reverse Transcription target specific primer s (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-IPNV-
Supplier: PRIMERDESIGN