ContactUsLinkComponent
qPCR detection of Mycobacterium avium subspecies paratuberculosis. Contains: Primer and probe, RT target specific primers (RNA virus es only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-MAP
Supplier:  PRIMERDESIGN
qPCR detection of Mycobacterium avium subspecies paratuberculosis. Contains: Primer and probe, RT target specific primers (RNA virus es only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-MAP
Supplier:  PRIMERDESIGN