ContactUsLinkComponent
qPCR detection of Budgerigar Fledgling Disease Virus (avian polyomavirus). Contains: Primer and probe, RT target specific primers (R NA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-APV
Supplier:  PRIMERDESIGN
qPCR detection of Budgerigar Fledgling Disease Virus (avian polyomavirus). Contains: Primer and probe, RT target specific primers (R NA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-APV
Supplier:  PRIMERDESIGN