ContactUsLinkComponent
qPCR detection of Grass Carp Reovirus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, En dogenous control, extraction control 1 * 150 Tests
Catalog # PATH-GCRV
Supplier:  PRIMERDESIGN
qPCR detection of Grass Carp Reovirus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, En dogenous control, extraction control 1 * 150 Tests
Catalog # PATH-GCRV
Supplier:  PRIMERDESIGN