qPCR detection of Viral Hemorrhagic Septicemia Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-VHSV-
Supplier: PRIMERDESIGN