qPCR detection of Human Parainfluenza virus type 3. Contains: Primer and probe, RT target specific primers (RNA viruses only), Stand ard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-HPIV3
Supplier: PRIMERDESIGN