ContactUsLinkComponent
Interior socket (can only be ordered with limited temperature-range - max. +7ß °C) ampacity 230 V, 2.2 A, can be switched off with t he ON/OFF switch, cannot be switched individually, moisture tight IP68 (option A8 necessary - see page 26) 1 * 1 items
Catalog # MEMMUF1060R3
Supplier:  MEMMERT
Interior socket (can only be ordered with limited temperature-range - max. +7ß °C) ampacity 230 V, 2.2 A, can be switched off with t he ON/OFF switch, cannot be switched individually, moisture tight IP68 (option A8 necessary - see page 26) 1 * 1 items
Catalog # MEMMUF1060R3
Supplier:  MEMMERT
Frequently Bought Together