ContactUsLinkComponent
HCC-1 Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 72 amino acids and having a mo lecular mass of 8411 Dalton. The HCC-1 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80136-10
Supplier:  Biorbyt
HCC-1 Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 72 amino acids and having a mo lecular mass of 8411 Dalton. The HCC-1 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80136-10
Supplier:  Biorbyt