ContactUsLinkComponent
GRO-Gamma Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 73 amino acids and having a molecular mass of 7902 Dalton. The CXCL3 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80135-10
Supplier:  Biorbyt
GRO-Gamma Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 73 amino acids and having a molecular mass of 7902 Dalton. The CXCL3 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80135-10
Supplier:  Biorbyt