GRO-Beta Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 73 amino acids and having a molecular mass of 7908 Dalton. The CXCL2 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80131-10
Supplier: Biorbyt