Exodus 2 Mouse Recombinant produced in E.coli as single, non-glycosylated, polypeptide chain containing 110 amino acids and having a molecular mass of 12 kDa. The CCL21 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80127-20
Supplier: Biorbyt